Assay Details
| Gene Symbol: LOC4330310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767520.2 | XP_015623006.1 | 9 - 10 | 2227 | 63 | |
| XM_015767521.2 | XP_015623007.1 | 9 - 10 | 2221 | 63 | ||
| XM_015767522.2 | XP_015623008.1 | 8 - 9 | 1939 | 63 | ||
| XM_015767523.2 | XP_015623009.1 | 9 - 10 | 2224 | 63 | ||
| XM_015767524.2 | XP_015623010.1 | 5 - 6 | 1517 | 63 | ||
| GenBank mRNA | AK241359.1 | - | 9 - 10 | 2224 | 63 | |
More Information
Set Membership: |
|

LOC4330310
Genomic Map