Assay Details
| Gene Symbol: LOC4330310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767520.2 | XP_015623006.1 | 10 - 11 | 2358 | 72 | |
| XM_015767521.2 | XP_015623007.1 | 10 - 11 | 2352 | 72 | ||
| XM_015767522.2 | XP_015623008.1 | 9 - 10 | 2070 | 72 | ||
| XM_015767523.2 | XP_015623009.1 | 10 - 11 | 2355 | 72 | ||
| XM_015767524.2 | XP_015623010.1 | 6 - 7 | 1648 | 72 | ||
| GenBank mRNA | AK241359.1 | - | 10 - 11 | 2355 | 72 | |
More Information
Set Membership: |
|

LOC4330310
Genomic Map