Assay Details
| Gene Symbol: LOC4330491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772121.2 | XP_015627607.1 | 11 - 12 | 2196 | 61 | |
| XM_015772122.2 | XP_015627608.1 | 10 - 11 | 1716 | 61 | ||
| GenBank mRNA | AK058346.1 | - | 1 - 2 | 61 | 61 | |
More Information
Set Membership: |
|

LOC4330491
Genomic Map