Assay Details
| Gene Symbol: LOC4330491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772121.2 | XP_015627607.1 | 16 - 17 | 3306 | 116 | |
| XM_015772122.2 | XP_015627608.1 | 15 - 16 | 2826 | 116 | ||
| GenBank mRNA | AK058346.1 | - | 6 - 7 | 1171 | 116 | |
More Information
Set Membership: |
|

LOC4330491
Genomic Map