Assay Details
| Gene Symbol: LOC4330593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772072.2 | XP_015627558.1 | 7 - 8 | 2703 | 86 | |
| XM_015772073.2 | XP_015627559.1 | 7 - 8 | 2207 | 86 | ||
| XM_015772074.2 | XP_015627560.1 | 7 - 8 | 3853 | 86 | ||
| GenBank mRNA | AK068228.1 | - | 4 - 5 | 666 | 86 | |
More Information
Set Membership: |
|

LOC4330593
Genomic Map