Assay Details
| Gene Symbol: LOC4330593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772072.2 | XP_015627558.1 | 8 - 9 | 2738 | 130 | |
| XM_015772073.2 | XP_015627559.1 | 8 - 9 | 2242 | 130 | ||
| XM_015772074.2 | XP_015627560.1 | 8 - 9 | 3888 | 130 | ||
| GenBank mRNA | AK068228.1 | - | 5 - 6 | 701 | 130 | |
More Information
Set Membership: |
|

LOC4330593
Genomic Map