Assay Details
| Gene Symbol: LOC4330593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772072.2 | XP_015627558.1 | 9 - 10 | 2911 | 98 | |
| XM_015772073.2 | XP_015627559.1 | 9 - 10 | 2415 | 98 | ||
| XM_015772074.2 | XP_015627560.1 | 9 - 10 | 4061 | 98 | ||
| GenBank mRNA | AK068228.1 | - | 6 - 7 | 874 | 98 | |
More Information
Set Membership: |
|

LOC4330593
Genomic Map