Assay Details
| Gene Symbol: LOC4330693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769669.2 | XP_015625155.1 | 2 - 3 | 432 | 65 | |
| GenBank mRNA | AK059970.1 | - | 2 - 3 | 332 | 65 | |
| AK069345.1 | - | 2 - 3 | 437 | 65 | ||
More Information
Set Membership: |
|

LOC4330693
Genomic Map