Assay Details
| Gene Symbol: LOC4330693 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769669.2 | XP_015625155.1 | 3 - 4 | 542 | 69 | |
| GenBank mRNA | AK059970.1 | - | 3 - 4 | 442 | 69 | |
| AK069345.1 | - | 3 - 4 | 547 | 69 | ||
More Information
Set Membership: |
|

LOC4330693
Genomic Map