Assay Details
| Gene Symbol: LOC4339877 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK318560.1 | - | 1848 | 132 | ||
| Gene Symbol: LOC4331752 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772500.2 | XP_015627986.1 | 3 - 4 | 707 | 132 | |
| GenBank mRNA | AK099530.1 | - | 3 - 4 | 691 | 132 | |
More Information
Set Membership: |
|

LOC4339877
Genomic Map