Assay Details
| Gene Symbol: LOC4331821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777807.2 | XP_015633293.1 | 6 - 7 | 1364 | 78 | |
| XM_015777808.2 | XP_015633294.1 | 6 - 7 | 1364 | 78 | ||
More Information
Set Membership: |
|

LOC4331821
Genomic Map