Assay Details
| Gene Symbol: LOC4331828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774864.2 | XP_015630350.1 | 2 - 3 | 505 | 88 | |
| XM_015774865.2 | XP_015630351.1 | 3 - 4 | 385 | 88 | ||
| GenBank mRNA | AB026558.1 | - | 3 - 4 | 331 | 88 | |
| AK059929.1 | - | 3 - 4 | 366 | 88 | ||
| AK060026.1 | - | 3 - 4 | 353 | 88 | ||
| AK101031.1 | - | 3 - 4 | 365 | 88 | ||
| AK104324.1 | - | 3 - 4 | 353 | 88 | ||
More Information
Set Membership: |
|

LOC4331828
Genomic Map