Assay Details
| Gene Symbol: LOC4331828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774864.2 | XP_015630350.1 | 5 - 6 | 742 | 105 | |
| XM_015774865.2 | XP_015630351.1 | 6 - 7 | 622 | 105 | ||
| GenBank mRNA | AB026558.1 | - | 6 - 7 | 568 | 105 | |
| AK059929.1 | - | 6 - 7 | 603 | 105 | ||
| AK060026.1 | - | 6 - 7 | 590 | 105 | ||
| AK101031.1 | - | 6 - 7 | 602 | 105 | ||
| AK104324.1 | - | 6 - 7 | 590 | 105 | ||
More Information
Set Membership: |
|

LOC4331828
Genomic Map