Assay Details
| Gene Symbol: LOC4331843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776789.2 | XP_015632275.1 | 1 - 2 | 360 | 150 | |
| GenBank mRNA | AK073133.1 | - | 1 - 2 | 300 | 150 | |
| AY323478.1 | - | 1 - 2 | 326 | 150 | ||
More Information
Set Membership: |
|

LOC4331843
Genomic Map