Assay Details
| Gene Symbol: LOC4332109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776559.1 | XP_015632045.1 | 14 - 15 | 1666 | 126 | |
| XM_015776560.1 | XP_015632046.1 | 14 - 15 | 1663 | 126 | ||
| GenBank mRNA | AK068416.1 | - | 14 - 15 | 1635 | 126 | |
More Information
Set Membership: |
|

LOC4332109
Genomic Map