Assay Details
| Gene Symbol: LOC4332109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776559.1 | XP_015632045.1 | 5 - 6 | 843 | 94 | |
| XM_015776560.1 | XP_015632046.1 | 5 - 6 | 840 | 94 | ||
| GenBank mRNA | AK068416.1 | - | 5 - 6 | 812 | 94 | |
More Information
Set Membership: |
|

LOC4332109
Genomic Map