Assay Details
Gene Symbol: LOC4332217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777431.2 | XP_015632917.1 | 5 - 6 | 992 | 71 | |
GenBank mRNA | AK071529.1 | - | 5 - 6 | 873 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
