Assay Details
Gene Symbol: LOC4332217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777431.2 | XP_015632917.1 | 6 - 7 | 1120 | 88 | |
GenBank mRNA | AK071529.1 | - | 6 - 7 | 1001 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
