Assay Details
| Gene Symbol: LOC4332324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772745.2 | XP_015628231.1 | 1 - 2 | 327 | 78 | |
| XM_015772746.2 | XP_015628232.1 | 1 - 2 | 327 | 78 | ||
| GenBank mRNA | AK067490.1 | - | 1 - 2 | 325 | 78 | |
More Information
Set Membership: |
|

LOC4332324
Genomic Map