Assay Details
| Gene Symbol: LOC4332424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776625.1 | XP_015632111.1 | 1 - 2 | 476 | 93 | |
| GenBank mRNA | AK060031.1 | - | 1 - 2 | 93 | 93 | |
| AK100908.1 | - | 1 - 2 | 482 | 93 | ||
| JX134102.1 | - | 1 - 2 | 202 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015776625.1 transcripts.
Set Membership: |
|

LOC4332424
Genomic Map