Assay Details
| Gene Symbol: LOC4332424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776625.1 | XP_015632111.1 | 3 - 4 | 660 | 92 | |
| GenBank mRNA | AB079064.1 | - | 2 - 3 | 178 | 92 | |
| AK060031.1 | - | 3 - 4 | 277 | 92 | ||
| AK100908.1 | - | 3 - 4 | 666 | 92 | ||
| JX134102.1 | - | 3 - 4 | 386 | 92 | ||
More Information
Set Membership: |
|

LOC4332424
Genomic Map