Assay Details
| Gene Symbol: LOC4332424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776625.1 | XP_015632111.1 | 6 - 7 | 902 | 76 | |
| GenBank mRNA | AB079064.1 | - | 5 - 6 | 420 | 76 | |
| AK060031.1 | - | 6 - 7 | 519 | 76 | ||
| AK100908.1 | - | 6 - 7 | 908 | 76 | ||
| JX134102.1 | - | 6 - 7 | 628 | 76 | ||
More Information
Set Membership: |
|

LOC4332424
Genomic Map