Assay Details
| Gene Symbol: LOC4332533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775963.2 | XP_015631449.1 | 1 - 2 | 1193 | 77 | |
| GenBank mRNA | AK072429.1 | - | 1 - 2 | 872 | 77 | |
| AK102156.1 | - | 1 - 2 | 1194 | 77 | ||
More Information
Set Membership: |
|

LOC4332533
Genomic Map