Assay Details
| Gene Symbol: LOC4332533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775963.2 | XP_015631449.1 | 2 - 3 | 2302 | 141 | |
| GenBank mRNA | AK072429.1 | - | 2 - 3 | 1981 | 141 | |
| AK102156.1 | - | 2 - 3 | 2303 | 141 | ||
More Information
Set Membership: |
|

LOC4332533
Genomic Map