Assay Details
| Gene Symbol: LOC4332552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772807.2 | XP_015628293.1 | 1 - 2 | 429 | 86 | |
| GenBank mRNA | AK061180.1 | - | 1 - 2 | 422 | 86 | |
| AK099089.1 | - | 1 - 2 | 398 | 86 | ||
More Information
Set Membership: |
|

LOC4332552
Genomic Map