Assay Details
| Gene Symbol: LOC4332552 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772807.2 | XP_015628293.1 | 2 - 3 | 566 | 62 | |
| GenBank mRNA | AK061180.1 | - | 2 - 3 | 559 | 62 | |
| AK099089.1 | - | 2 - 3 | 535 | 62 | ||
More Information
Set Membership: |
|

LOC4332552
Genomic Map