Assay Details
| Gene Symbol: LOC4332692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776931.2 | XP_015632417.1 | 3 - 4 | 523 | 102 | |
| XM_015776932.2 | XP_015632418.1 | 3 - 4 | 523 | 102 | ||
| XM_015776933.2 | XP_015632419.1 | 3 - 4 | 523 | 102 | ||
| GenBank mRNA | AK066439.1 | - | 3 - 4 | 518 | 102 | |
More Information
Set Membership: |
|

LOC4332692
Genomic Map