Assay Details
| Gene Symbol: LOC4332812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776549.2 | 8 - 9 | 1314 | 73 | ||
| GenBank mRNA | AK065579.1 | - | 10 - 11 | 1217 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4332812| Gene Symbol: LOC4332812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776549.2 | 8 - 9 | 1314 | 73 | ||
| GenBank mRNA | AK065579.1 | - | 10 - 11 | 1217 | 73 | |
Set Membership: |
|