Assay Details
| Gene Symbol: LOC4332812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776549.2 | 4 - 5 | 836 | 110 | ||
| XM_015776550.2 | XP_015632036.1 | 4 - 5 | 836 | 110 | ||
| GenBank mRNA | AK065579.1 | - | 6 - 7 | 739 | 110 | |
More Information
Set Membership: |
|

LOC4332812
Genomic Map