Assay Details
| Gene Symbol: LOC4332812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776549.2 | 5 - 6 | 906 | 152 | ||
| XM_015776550.2 | XP_015632036.1 | 5 - 6 | 906 | 152 | ||
| GenBank mRNA | AK065579.1 | - | 7 - 8 | 809 | 152 | |
More Information
Set Membership: |
|

LOC4332812
Genomic Map