Assay Details
| Gene Symbol: LOC4332903 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776859.2 | XP_015632345.1 | 1 - 2 | 319 | 67 | |
| GenBank mRNA | AK103387.1 | - | 1 - 2 | 307 | 67 | |
More Information
Set Membership: |
|

LOC4332903
Genomic Map