Assay Details
| Gene Symbol: LOC4332904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777208.2 | XP_015632694.1 | 8 - 9 | 948 | 112 | |
| GenBank mRNA | AK066313.1 | - | 8 - 9 | 946 | 112 | |
| AK066352.1 | - | 8 - 9 | 901 | 112 | ||
More Information
Set Membership: |
|

LOC4332904
Genomic Map