Assay Details
| Gene Symbol: LOC4333424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773656.2 | XP_015629142.1 | 2 - 3 | 1732 | 64 | |
| GenBank mRNA | AK102418.1 | - | 2 - 3 | 469 | 64 | |
More Information
Set Membership: |
|

LOC4333424
Genomic Map