Assay Details
| Gene Symbol: LOC4333500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777232.2 | XP_015632718.1 | 11 - 12 | 1461 | 103 | |
| XM_015777233.2 | XP_015632719.1 | 11 - 12 | 1466 | 103 | ||
| XM_015777234.2 | XP_015632720.1 | 10 - 11 | 1387 | 103 | ||
| XM_015777235.2 | XP_015632721.1 | 10 - 11 | 1298 | 103 | ||
| XM_015777236.2 | XP_015632722.1 | 10 - 11 | 1392 | 103 | ||
| XM_015777238.2 | XP_015632724.1 | 10 - 11 | 1303 | 103 | ||
| XM_015777239.2 | XP_015632725.1 | 11 - 12 | 1365 | 103 | ||
| GenBank mRNA | AK066665.1 | - | 10 - 11 | 1154 | 103 | |
More Information
Set Membership: |
|

LOC4333500
Genomic Map