Assay Details
| Gene Symbol: LOC4333500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777232.2 | XP_015632718.1 | 13 - 14 | 1633 | 69 | |
| XM_015777233.2 | XP_015632719.1 | 13 - 14 | 1638 | 69 | ||
| XM_015777234.2 | XP_015632720.1 | 12 - 13 | 1559 | 69 | ||
| XM_015777235.2 | XP_015632721.1 | 12 - 13 | 1470 | 69 | ||
| XM_015777236.2 | XP_015632722.1 | 12 - 13 | 1564 | 69 | ||
| XM_015777238.2 | XP_015632724.1 | 12 - 13 | 1475 | 69 | ||
| XM_015777239.2 | XP_015632725.1 | 13 - 14 | 1537 | 69 | ||
| GenBank mRNA | AK066665.1 | - | 12 - 13 | 1326 | 69 | |
More Information
Set Membership: |
|

LOC4333500
Genomic Map