Assay Details
| Gene Symbol: LOC4333500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777232.2 | XP_015632718.1 | 1 - 2 | 222 | 62 | |
| XM_015777234.2 | XP_015632720.1 | 1 - 2 | 220 | 62 | ||
| XM_015777235.2 | XP_015632721.1 | 1 - 2 | 220 | 62 | ||
| XM_015777236.2 | XP_015632722.1 | 1 - 2 | 220 | 62 | ||
| XM_015777238.2 | XP_015632724.1 | 1 - 2 | 220 | 62 | ||
| XM_015777239.2 | XP_015632725.1 | 1 - 2 | 220 | 62 | ||
| GenBank mRNA | AK066665.1 | - | 1 - 2 | 76 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777232.2, XM_015777234.2, XM_015777235.2, XM_015777236.2, XM_015777238.2, XM_015777239.2 transcripts.
Set Membership: |
|

LOC4333500
Genomic Map