Assay Details
| Gene Symbol: LOC4333500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777235.2 | XP_015632721.1 | 3 - 4 | 365 | 76 | |
| GenBank mRNA | AK066665.1 | - | 3 - 4 | 221 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777235.2 transcripts.
Set Membership: |
|

LOC4333500
Genomic Map