Assay Details
| Gene Symbol: LOC4333500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777232.2 | XP_015632718.1 | 5 - 6 | 668 | 110 | |
| XM_015777233.2 | XP_015632719.1 | 5 - 6 | 673 | 110 | ||
| XM_015777234.2 | XP_015632720.1 | 4 - 5 | 594 | 110 | ||
| XM_015777235.2 | XP_015632721.1 | 4 - 5 | 505 | 110 | ||
| XM_015777236.2 | XP_015632722.1 | 4 - 5 | 599 | 110 | ||
| XM_015777238.2 | XP_015632724.1 | 4 - 5 | 510 | 110 | ||
| XM_015777239.2 | XP_015632725.1 | 5 - 6 | 572 | 110 | ||
| GenBank mRNA | AK066665.1 | - | 4 - 5 | 361 | 110 | |
More Information
Set Membership: |
|

LOC4333500
Genomic Map