Assay Details
| Gene Symbol: LOC4333500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777232.2 | XP_015632718.1 | 9 - 10 | 1129 | 86 | |
| XM_015777233.2 | XP_015632719.1 | 9 - 10 | 1134 | 86 | ||
| XM_015777234.2 | XP_015632720.1 | 8 - 9 | 1055 | 86 | ||
| XM_015777235.2 | XP_015632721.1 | 8 - 9 | 966 | 86 | ||
| XM_015777236.2 | XP_015632722.1 | 8 - 9 | 1060 | 86 | ||
| XM_015777238.2 | XP_015632724.1 | 8 - 9 | 971 | 86 | ||
| XM_015777239.2 | XP_015632725.1 | 9 - 10 | 1033 | 86 | ||
More Information
Set Membership: |
|

LOC4333500
Genomic Map