Assay Details
| Gene Symbol: LOC4333569 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776051.2 | XP_015631537.1 | 2 - 3 | 946 | 77 | |
| GenBank mRNA | AK069229.1 | - | 2 - 3 | 947 | 77 | |
More Information
Set Membership: |
|

LOC4333569
Genomic Map