Assay Details
| Gene Symbol: LOC4333918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774831.2 | XP_015630317.1 | 3 - 4 | 459 | 152 | |
| GenBank mRNA | AB116722.1 | - | 3 - 4 | 335 | 152 | |
| AK105178.1 | - | 3 - 4 | 390 | 152 | ||
More Information
Set Membership: |
|

LOC4333918
Genomic Map