Assay Details
| Gene Symbol: LOC4333919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB047313.1 | - | 1 - 2 | 84 | 115 | |
| AK058421.1 | - | 92 | 115 | |||
| KC140126.1 | - | 46 | 115 | |||
| KX302608.1 | - | 75 | 115 | |||
More Information
Set Membership: |
|

LOC4333919
Genomic Map