Assay Details
| Gene Symbol: LOC4333922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773436.2 | XP_015628922.1 | 1 - 2 | 412 | 72 | |
| GenBank mRNA | EF431929.1 | - | 1 - 2 | 324 | 72 | |
More Information
Set Membership: |
|

LOC4333922
Genomic Map