Assay Details
| Gene Symbol: LOC4333922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773436.2 | XP_015628922.1 | 3 - 4 | 533 | 122 | |
| GenBank mRNA | EF431929.1 | - | 3 - 4 | 445 | 122 | |
More Information
Set Membership: |
|

LOC4333922
Genomic Map