Assay Details
| Gene Symbol: LOC4333922 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773436.2 | XP_015628922.1 | 4 - 5 | 644 | 105 | |
| GenBank mRNA | EF431929.1 | - | 4 - 5 | 556 | 105 | |
More Information
Set Membership: |
|

LOC4333922
Genomic Map