Assay Details
| Gene Symbol: LOC4333976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773759.2 | XP_015629245.1 | 2 - 3 | 484 | 54 | |
| GenBank mRNA | AK071011.1 | - | 2 - 3 | 479 | 54 | |
More Information
Set Membership: |
|

LOC4333976
Genomic Map