Assay Details
| Gene Symbol: LOC4334040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772664.2 | XP_015628150.1 | 14 - 15 | 1308 | 74 | |
| XM_015772665.2 | XP_015628151.1 | 14 - 15 | 1333 | 74 | ||
| GenBank mRNA | AK105468.1 | - | 14 - 15 | 1302 | 74 | |
More Information
Set Membership: |
|

LOC4334040
Genomic Map