Assay Details
| Gene Symbol: LOC4334040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772664.2 | XP_015628150.1 | 1 - 2 | 323 | 139 | |
| XM_015772665.2 | XP_015628151.1 | 1 - 2 | 325 | 139 | ||
| GenBank mRNA | AK101854.1 | - | 1 - 2 | 316 | 139 | |
| AK105468.1 | - | 1 - 2 | 317 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772664.2, XM_015772665.2 transcripts.
Set Membership: |
|

LOC4334040
Genomic Map