Assay Details
| Gene Symbol: LOC4334040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772664.2 | XP_015628150.1 | 8 - 9 | 866 | 101 | |
| XM_015772665.2 | XP_015628151.1 | 8 - 9 | 868 | 101 | ||
| GenBank mRNA | AK101854.1 | - | 8 - 9 | 859 | 101 | |
| AK105468.1 | - | 8 - 9 | 860 | 101 | ||
More Information
Set Membership: |
|

LOC4334040
Genomic Map