Assay Details
| Gene Symbol: LOC4334067 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777643.2 | XP_015633129.1 | 4 - 5 | 803 | 97 | |
| GenBank mRNA | AK060249.1 | - | 1 - 2 | 121 | 97 | |
More Information
Set Membership: |
|

LOC4334067
Genomic Map